Thymine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil

T0234-25G T0234-5G T0234-100G
TCIAT0234-25GEA 340 NOK
TCIAT0234-25G TCIAT0234-5G TCIAT0234-100G
Thymine ≥98.0% (by HPLC, titration analysis)
Thymine
Formel: C₅H₆N₂O₂
MW: 126,11 g/mol
Kokepunkt: 389 °C (1013 hPa)
Smeltepunkt: 320°C
Tetthet: 1,454 g/cm³ (25 °C)
MDL nummer: MFCD00006026
CAS-nummer: 65-71-4
EINECS: 200-616-1
Merck Indeks: 14,09398
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f73c58f3b2d92f6ff38c490d38be6be9c76bed&vid=17f73c58f3b2d92f6ff38c490d38be6be9c76bed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR