Methyl-2,5-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester
M2006-25G M2006-5G
TCIAM2006-25GEA 1150 NOK
TCIAM2006-25G TCIAM2006-5G
Methyl-2,5-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl-2,5-dihydroxybenzoate
Formel: C₈H₈O₄
MW: 168,15 g/mol
Kokepunkt: 136°C
MDL nummer: MFCD00016464
CAS-nummer: 2150-46-1
EINECS: 218-427-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37453e90be2047c9dc57e833625da1b7f0e0d2e6&vid=37453e90be2047c9dc57e833625da1b7f0e0d2e6&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C