2-Amino-4-methylpyrimidine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine

A1474-25G A1474-5G
TCIAA1474-25GEA 1300 NOK
TCIAA1474-25G TCIAA1474-5G
2-Amino-4-methylpyrimidine ≥97.0% (by titrimetric analysis)
2-Amino-4-methylpyrimidine
Formel: C₅H₇N₃
MW: 109,13 g/mol
Kokepunkt: 283 °C (1013 hPa)
Smeltepunkt: 155…157 °C
Tetthet: 1 g/cm³ (20 °C)
MDL nummer: MFCD00006101
CAS-nummer: 108-52-1
EINECS: 203-591-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a66cc79dffc1d3cbc558c117afc40facb7427d0&vid=3a66cc79dffc1d3cbc558c117afc40facb7427d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 156.0 to 161.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR