2-Amino-6-methoxybenzothiazole ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 6-Methoxybenzothiazol-2-ylamine

A0715-25G A0715-500G A0715-100G
TCIAA0715-25GEA 260 NOK
TCIAA0715-25G TCIAA0715-500G TCIAA0715-100G
2-Amino-6-methoxybenzothiazole ≥98.0% (by titrimetric analysis)
2-Amino-6-methoxybenzothiazole
Formel: C₈H₈N₂OS
MW: 180,23 g/mol
Kokepunkt: 348 °C (1013 hPa)
Smeltepunkt: 160 °C
MDL nummer: MFCD00005787
CAS-nummer: 1747-60-0
EINECS: 217-130-0
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2b8e1f4a1a93bc385db581ad857baaff6b7fcb5&vid=d2b8e1f4a1a93bc385db581ad857baaff6b7fcb5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to lump
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol very faint turbidity
Melting point 165.0 to 170.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR