2-Bromo-5-fluorobenzaldehyde ≥95.0% (by GC)

Leverandør: TCI
Warning

B2353-25G B2353-5G
TCIAB2353-25GEA 1850 NOK
TCIAB2353-25G TCIAB2353-5G
2-Bromo-5-fluorobenzaldehyde ≥95.0% (by GC)
2-Bromo-5-fluorobenzaldehyde
Formel: C₇H₄BrFO
MW: 203,01 g/mol
MDL nummer: MFCD00142872
CAS-nummer: 94569-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55991607aebee2b41b2e17844b100db439b2f83&vid=b55991607aebee2b41b2e17844b100db439b2f83&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 53.0 to 57.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR