6-Fluoro-m-anisaldehyde ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Fluoro-5-methoxybenzaldehyde, 6-Fluoro-3-anisaldehyde
F0383-25G
F0383-5G
TCIAF0383-25GEA
6000
NOK
TCIAF0383-25G
TCIAF0383-5G
6-Fluoro-m-anisaldehyde ≥95.0% (by GC)
6-Fluoro-m-anisaldehyde
Formel:
C₈H₇FO₂ MW: 154,14 g/mol |
MDL nummer:
MFCD00070795 CAS-nummer: 105728-90-3 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...