3,3,3-Trifluoropropionic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
3,3,3-Trifluoropropanoic acid
T1713-1G
T1713-5G
TCIAT1713-1GEA
285
NOK
TCIAT1713-1G
TCIAT1713-5G
3,3,3-Trifluoropropionic acid ≥98.0% (by GC, titration analysis)
3,3,3-Trifluoropropionic acid
Formel:
C₃H₃F₃O₂ MW: 128,05 g/mol Kokepunkt: 153°C Smeltepunkt: 0 °C Tetthet: 1,441 g/cm³ (27 °C) |
MDL nummer:
MFCD00153292 CAS-nummer: 2516-99-6 FN: 2924 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...