3-Acetylindole ≥98.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: 3-Indolyl methyl ketone, 1-(3-Indolyl)-1-ethanone, 1-(1H-Indol-3-yl)ethanone, Indol-3-yl methyl ketone

A0849-25G A0849-5G
TCIAA0849-25GEA 1575 NOK
TCIAA0849-25G TCIAA0849-5G
3-Acetylindole ≥98.0% (by total nitrogen basis)
3-Acetylindole
Formel: C₁₀H₉NO
MW: 159,19 g/mol
Kokepunkt: 144 - 145 °C (10 -rr)
Smeltepunkt: 186…189 °C
MDL nummer: MFCD00005626
CAS-nummer: 703-80-0
EINECS: 211-875-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c30db85fa5e92eca150d685fec048a703cf32417&vid=c30db85fa5e92eca150d685fec048a703cf32417&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light red to Green powder to crystalline
Purity (with Total Nitrogen) min. 98.0 %
Melting point 189.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR