Indole-3-carbaldehyd ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

I0027-25G I0027-5G
TCIAI0027-25GEA 905 NOK
TCIAI0027-25G TCIAI0027-5G
Indole-3-carbaldehyd ≥98.0% (by GC)
Indole-3-carbaldehyd
Formel: C₉H₇NO
MW: 145,16 g/mol
Smeltepunkt: 192…195 °C
Tetthet: 1,34 g/cm³ (20 °C)
MDL nummer: MFCD00005622
CAS-nummer: 487-89-8
EINECS: 207-665-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05fe62a04c57b3414f943f999aca8b799a5d4572&vid=05fe62a04c57b3414f943f999aca8b799a5d4572&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 200.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR