4-tert-Butylbenzenesulphonyl chloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 4-tert-Butylbenzene-1-sulphonyl chloride

B1983-25G B1983-5G
TCIAB1983-25GEA 755 NOK
TCIAB1983-25G TCIAB1983-5G
4-tert-Butylbenzenesulphonyl chloride ≥98.0% (by titrimetric analysis)
4-tert-Butylbenzenesulphonyl chloride
Formel: C₁₀H₁₃ClO₂S
MW: 232,73 g/mol
Kokepunkt: 165°C
Smeltepunkt: 83 °C
MDL nummer: MFCD00007449
CAS-nummer: 15084-51-2
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR