4-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

A2383-1G A2383-5G
TCIAA2383-1GEA 905 NOK
TCIAA2383-1G TCIAA2383-5G
4-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-fluoropyridine
Formel: C₅H₅FN₂
MW: 112,11 g/mol
MDL nummer: MFCD00234072
CAS-nummer: 2247-88-3
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=038e8a4cf2a433f8fb94108f5e57aefabe22c4ce&vid=038e8a4cf2a433f8fb94108f5e57aefabe22c4ce&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR