5-Amino-2-methylindole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole

A1539-1G A1539-5G
TCIAA1539-1GEA 350 NOK
TCIAA1539-1G TCIAA1539-5G
5-Amino-2-methylindole ≥98.0% (by GC, titration analysis)
5-Amino-2-methylindole
Formel: C₉H₁₀N₂
MW: 146,19 g/mol
Smeltepunkt: 158…159 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02093426
CAS-nummer: 7570-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edde7b1b7b01cc6c913ed95d3edfa62ab620f3cb&vid=edde7b1b7b01cc6c913ed95d3edfa62ab620f3cb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark blue to Black powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR