5-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
Leverandør: TCI
|
Warning
|
A1395-1G
A1395-25G
A1395-5G
TCIAA1395-1GEA
220
NOK
TCIAA1395-1G
TCIAA1395-25G
TCIAA1395-5G
5-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
5-Amino-4,6-dichloropyrimidine
Formel:
C₄H₃Cl₂N₃ MW: 163,99 g/mol Kokepunkt: 262 °C (1013 hPa) Smeltepunkt: 147 °C |
MDL nummer:
MFCD00006108 CAS-nummer: 5413-85-4 EINECS: 226-503-7 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 143.0 to 147.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...