5-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

B2302-1G B2302-5G
TCIAB2302-1GEA 300 NOK
TCIAB2302-1G TCIAB2302-5G
5-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2-thenoic acid
Formel: C₅H₃BrO₂S
MW: 207,05 g/mol
Smeltepunkt: 142°C
MDL nummer: MFCD00079725
CAS-nummer: 7311-63-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR