5-Methylisoxazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

M1740-25G M1740-5G
TCIAM1740-25GEA 1300 NOK
TCIAM1740-25G TCIAM1740-5G
5-Methylisoxazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Methylisoxazole-4-carboxylic acid
Formel: C₅H₅NO₃
MW: 127,1 g/mol
MDL nummer: MFCD00955655
CAS-nummer: 42831-50-5
EINECS: 222-289-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ae4f67f8e5cc41dcd9dde7696c0718247eda5f9&vid=9ae4f67f8e5cc41dcd9dde7696c0718247eda5f9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR