9-Anthracenemethanol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 9-Anthrylmethanol, Anthracene-9-methanol

H0670-25G H0670-250G
TCIAH0670-25GEA 1275 NOK
TCIAH0670-25G TCIAH0670-250G
9-Anthracenemethanol ≥98.0% (by GC)
9-Anthracenemethanol
Formel: C₁₅H₁₂O
MW: 208,26 g/mol
Kokepunkt: 371 °C (1013 hPa)
Smeltepunkt: 162…164 °C
Tetthet: 1,3 g/cm³ (20 °C)
MDL nummer: MFCD00001264
CAS-nummer: 1468-95-7
EINECS: 215-998-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f739f8a4448ddee91d70e068e0615db2b71ee61&vid=4f739f8a4448ddee91d70e068e0615db2b71ee61&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 167.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR