Benzothiazole ≥96.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Benzo[d]thiazole

B0092-25ML B0092-500ML B0092-100ML
TCIAB0092-25MLEA 220 NOK
TCIAB0092-25ML TCIAB0092-500ML TCIAB0092-100ML
Benzothiazole ≥96.0% (by GC)
Benzothiazole
Formel: C₇H₅NS
MW: 135,19 g/mol
Kokepunkt: 231°C
Tetthet: 1,246 g/cm³ (20 °C)
Flash punkt: 107°C
MDL nummer: MFCD00005775
CAS-nummer: 95-16-9
EINECS: 202-396-2
FN: 2810
ADR: 6.1,III
Merck Indeks: 14,01107
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac384398827cfa1e792561d2ef78d4789cda7147&vid=ac384398827cfa1e792561d2ef78d4789cda7147&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR