Isopropyl chloroacetate ≥98.0% (by GC)

Leverandør: TCI
Danger

C0948-25ML C0948-500ML
TCIAC0948-25MLEA 220 NOK
TCIAC0948-25ML TCIAC0948-500ML
Isopropyl chloroacetate ≥98.0% (by GC)
Isopropyl chloroacetate
Formel: C₅H₉ClO₂
MW: 136,58 g/mol
Kokepunkt: 150°C
Smeltepunkt: –112 °C
Tetthet: 1,085 g/cm³ (20 °C)
Flash punkt: 45°C
MDL nummer: MFCD00040410
CAS-nummer: 105-48-6
EINECS: 203-301-7
FN: 2947
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29b4c4db5a14484f5216f30a7fcd433001ec8cec&vid=29b4c4db5a14484f5216f30a7fcd433001ec8cec&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR