N-Benzylmaleimide ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: 1-Benzyl-1H-pyrrole-2,5-dione, 1-Benzylpyrrole-2,5-dione

B1805-25G B1805-5G
TCIAB1805-25GEA 1650 NOK
TCIAB1805-25G TCIAB1805-5G
N-Benzylmaleimide ≥98.0% (by HPLC)
N-Benzylmaleimide
Formel: C₁₁H₉NO₂
MW: 187,2 g/mol
Kokepunkt: 177°C
Smeltepunkt: 70…72 °C
Tetthet: 1,296 g/cm³ (20 °C)
MDL nummer: MFCD00014540
CAS-nummer: 1631-26-1
EINECS: 216-631-1
FN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb489c1c8e7ec8eec4be824a3bb97ce396a17c55&vid=cb489c1c8e7ec8eec4be824a3bb97ce396a17c55&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR