Etylcyanoacetat ≥99.0% (by GC)

Leverandør: TCI
Warning

C0441-25G C0441-500G
TCIAC0441-25GEA 270 NOK
TCIAC0441-25G TCIAC0441-500G
Etylcyanoacetat ≥99.0% (by GC)
Etylcyanoacetat
Formel: C₅H₇NO₂
MW: 113,12 g/mol
Kokepunkt: 209 °C (1013 hPa)
Smeltepunkt: –22 °C
Tetthet: 1,063 g/cm³ (20 °C)
Flash punkt: 110°C
MDL nummer: MFCD00001940
CAS-nummer: 105-56-6
EINECS: 203-309-0
Merck Indeks: 14,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dbe76ab860a87e843e238db979e99be310bead7&vid=4dbe76ab860a87e843e238db979e99be310bead7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR