Bis(dimethylamino)dimethylsilane ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
B2150-25ML
B2150-250ML
TCIAB2150-25MLEA
395
NOK
TCIAB2150-25ML
TCIAB2150-250ML
Bis(dimethylamino)dimethylsilane ≥97.0% (by GC)
Bis(dimethylamino)dimethylsilane
Formel:
C₆H₁₈N₂Si MW: 146,31 g/mol Kokepunkt: 128°C Smeltepunkt: –98 °C Tetthet: 0,809 g/cm³ (22 °C) Flash punkt: -7°C |
MDL nummer:
MFCD00008296 CAS-nummer: 3768-58-9 EINECS: 223-200-1 FN: 2924 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...