Furfuryl acetate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: Furan-2-ylmethyl acetate, Acetic acid furfuryl ester

A0903-25ML A0903-100ML
TCIAA0903-25MLEA 405 NOK
TCIAA0903-25ML TCIAA0903-100ML
Furfuryl acetate ≥97.0% (by GC)
Furfuryl acetate
Formel: C₇H₈O₃
MW: 140,14 g/mol
Kokepunkt: 175 °C (1013 hPa)
Tetthet: 1,1176 g/cm³ (20 °C)
Flash punkt: 65°C
MDL nummer: MFCD00003251
CAS-nummer: 623-17-6
EINECS: 210-775-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe051d5dad9c16d50471d9b5f5a6e943c64df61f&vid=fe051d5dad9c16d50471d9b5f5a6e943c64df61f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR