2,6-Bis(hydroxymethyl)-p-cresol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Methyl-2,6-bis(hydroxymethyl)phenol, 1,3-Bis(hydroxymethyl)-2-hydroxy-5-methylbenzene, 2,6-Bis(hydroxymethyl)-4-methylphenol, 3,5-Bis(hydroxymethyl)-4-hydroxytoluene

B1525-25G B1525-250G
TCIAB1525-25GEA 660 NOK
TCIAB1525-25G TCIAB1525-250G
2,6-Bis(hydroxymethyl)-p-cresol ≥98.0% (by GC)
2,6-Bis(hydroxymethyl)-p-cresol
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Kokepunkt: 363 °C (1013 hPa)
Smeltepunkt: 133…134 °C
Tetthet: 1,382 g/cm³ (-173 °C)
MDL nummer: MFCD00004619
CAS-nummer: 91-04-3
EINECS: 202-036-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc0ae98fa2502539fbba6461e80e6950a074957c&vid=bc0ae98fa2502539fbba6461e80e6950a074957c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 128.0 deg-C
Solubility in THF very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR