δ-Valerolactone ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Pentan-5-olide, Tetrahydro-2H-pyran-2-one, 5-Hydroxypentanoic acid lactone, δ-Valerolactone
V0039-25G
V0039-500G
V0039-100G
TCIAV0039-25GEA
300
NOK
TCIAV0039-25G
TCIAV0039-500G
TCIAV0039-100G
δ-Valerolactone ≥98.0% (by GC)
δ-Valerolactone
Formel:
C₅H₈O₂ MW: 100,12 g/mol Kokepunkt: 220°C Smeltepunkt: –10 °C Tetthet: 1,0988 g/cm³ (20 °C) Flash punkt: 107°C Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00006645 CAS-nummer: 542-28-9 EINECS: 208-807-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid to slightly cloudy liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...