Butylene di(acetate) ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
1,4-Diacetoxybutane, Butylene glycol diacetate, Butane-1,4-diyl diacetate
B1232-25G
B1232-500G
TCIAB1232-25GEA
220
NOK
TCIAB1232-25G
TCIAB1232-500G
Butylene di(acetate) ≥98.0% (by GC)
Butylene di(acetate)
Formel:
C₈H₁₄O₄ MW: 174,2 g/mol Kokepunkt: 230°C |
MDL nummer:
MFCD00014984 CAS-nummer: 628-67-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...