Hexylene di(acetate) ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
1,6-Diacetoxyhexane, Hexylene glycol diacetate, Hexane-1,6-diyl diacetate, 1,6-Hexanediol diacetate
H0832-25ML
H0832-500ML
TCIAH0832-25MLEA
220
NOK
TCIAH0832-25ML
TCIAH0832-500ML
Hexylene di(acetate) ≥98.0% (by GC)
Hexylene di(acetate)
Formel:
C₁₀H₁₈O₄ MW: 202,25 g/mol Kokepunkt: 260°C |
MDL nummer:
MFCD00191655 CAS-nummer: 6222-17-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...