4-Aminohipposyre ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
p-Aminohippuric acid, 4-Aminohippuric Acid, 2-(4-Aminobenzamido)acetic acid, N-(4-Aminobenzoyl)glycine
A0313-10G
A0313-25G
TCIAA0313-10GEA
1750
NOK
TCIAA0313-10G
TCIAA0313-25G
4-Aminohipposyre ≥99.0% (by titrimetric analysis)
4-Aminohippuric acid
Formel:
H₂NC₆H₄C(O)NHCH₂COOH MW: 194,19 g/mol Kokepunkt: 441 °C (1013 hPa) Smeltepunkt: 199…202 °C Tetthet: 1,402 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007890 CAS-nummer: 61-78-9 EINECS: 200-518-9 Merck Indeks: 14,00442 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...