4-tert-Oktylfenol ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-tert-Octylphenol, 4-(1,1,3,3-Tetramethylbutyl)phenol

T0144-500G T0144-25G
TCIAT0144-500GEA 475 NOK
TCIAT0144-500G TCIAT0144-25G
4-tert-Oktylfenol ≥95.0% (by GC)
4-tert-Oktylfenol
Formel: C₁₄H₂₂O
MW: 206,33 g/mol
Kokepunkt: 279°C
Smeltepunkt: 80…81 °C
Tetthet: 0,932 g/cm³ (20 °C)
Flash punkt: 147 °C
MDL nummer: MFCD00002368
CAS-nummer: 140-66-9
EINECS: 205-426-2
FN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0088ed289b7060423c3ab7e4ffe269ea01fc2617&vid=0088ed289b7060423c3ab7e4ffe269ea01fc2617&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR