4-Amino-4H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine
A1137-25G
A1137-250G
TCIAA1137-25GEA
595
NOK
TCIAA1137-25G
TCIAA1137-250G
4-Amino-4H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)
4-Amino-4H-1,2,4-triazole
Formel:
C₂H₄N₄ MW: 84,08 g/mol |
MDL nummer:
MFCD00003099 CAS-nummer: 584-13-4 EINECS: 209-533-5 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...