5-Bromo-2-chlorophenol ≥98.0% (by GC)

Leverandør: TCI
Warning

B2919-1G B2919-5G
TCIAB2919-1GEA 445 NOK
TCIAB2919-1G TCIAB2919-5G
5-Bromo-2-chlorophenol ≥98.0% (by GC)
5-Bromo-2-chlorophenol
Formel: C₆H₄BrClO
MW: 207,45 g/mol
Kokepunkt: 234°C
MDL nummer: MFCD00672940
CAS-nummer: 183802-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d499ea2e93f4f21019628711d8bd0d055cc05df0&vid=d499ea2e93f4f21019628711d8bd0d055cc05df0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR