(±)-2-Heptanol ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol
H0034-25ML
H0034-500ML
H0034-100ML
TCIAH0034-25MLEA
230
NOK
TCIAH0034-25ML
TCIAH0034-500ML
TCIAH0034-100ML
(±)-2-Heptanol ≥98.0% (by GC)
(±)-2-Heptanol
Formel:
C₇H₁₆O MW: 116,2 g/mol Kokepunkt: 160°C Tetthet: 0,82 g/cm³ (20 °C) Flash punkt: 64°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004587 CAS-nummer: 543-49-7 EINECS: 208-844-3 ADR: 3,III Merck Indeks: 14,04662 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...