(R)-(+)-Limonen ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene
L0105-25ML
L0105-5ML
TCIAL0105-25MLEA
2225
NOK
TCIAL0105-25ML
TCIAL0105-5ML
(R)-(+)-Limonen ≥99.0% (by GC)
(R)-(+)-Limonen
Formel:
C₁₀H₁₆ MW: 136,24 g/mol Kokepunkt: 176°C Smeltepunkt: –75°C Tetthet: 0,8411 g/cm³ (20 °C) Flash punkt: 51°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00062991 CAS-nummer: 5989-27-5 EINECS: 227-813-5 FN: 2319 ADR: 3,III Merck Indeks: 14,05493 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Specific rotation [a]20/D | +115.0 to +125.0 deg (neat) |