(S)-(-)-Limonene ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
L0132-25ML
L0132-5ML
L0132-500ML
TCIAL0132-25MLEA
340
NOK
TCIAL0132-25ML
TCIAL0132-5ML
TCIAL0132-500ML
(S)-(-)-Limonene ≥95.0% (by GC)
(S)-(-)-Limonene
Formel:
C₁₀H₁₆ MW: 136,24 g/mol Kokepunkt: 64°C Smeltepunkt: –74 °C Tetthet: 0,84 g/cm³ (20 °C) Flash punkt: 45°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00001558 CAS-nummer: 5989-54-8 EINECS: 227-815-6 FN: 2319 ADR: 3,III Merck Indeks: 14,05493 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Optical purity (GC) | min. 96.0 ee% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...