1,10-Dibromdekan ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
1,10-Dibromodecan, Decane, 1,10-dibromo-
D0179-25G
D0179-100G
TCIAD0179-25GEA
340
NOK
TCIAD0179-25G
TCIAD0179-100G
1,10-Dibromdekan ≥95.0% (by GC)
1,10-Dibromdekan
Formel:
BrH₂C(CH₂)₈CH₂Br MW: 300,08 g/mol Kokepunkt: 298 °C (1013 hPa) Smeltepunkt: 27 °C Tetthet: 1,35 g/cm³ (25 °C) Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00000222 CAS-nummer: 4101-68-2 EINECS: 223-871-0 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...