1,12-Dibromdodekan ≥98.0% (by GC)
Leverandør: TCI
D1346-100G
D1346-25G
D1346-500G
TCIAD1346-100GEA
2075
NOK
TCIAD1346-100G
TCIAD1346-25G
TCIAD1346-500G
1,12-Dibromdodekan ≥98.0% (by GC)
1,12-Dibromdodekan
Formel:
BrH₂C(CH₂)₁₀CH₂Br MW: 328,13 g/mol Kokepunkt: 215°C Smeltepunkt: 37…38 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00000226 CAS-nummer: 3344-70-5 EINECS: 222-096-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...