Bromacetaldehyd dietyl acetal ≥95.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
2-Bromo-1,1-diethoxyethane
B0957-25ML
B0957-500ML
B0957-100ML
TCIAB0957-25MLEA
220
NOK
TCIAB0957-25ML
TCIAB0957-500ML
TCIAB0957-100ML
Bromacetaldehyd dietyl acetal ≥95.0% (by GC)
Bromacetaldehyd dietyl acetal
Formel:
BrH₂CCH(OCH₂CH₃)₂ MW: 197,07 g/mol Kokepunkt: 67°C Tetthet: 1,27 g/cm³ (20 °C) Flash punkt: 51°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00000214 CAS-nummer: 2032-35-1 EINECS: 217-989-1 FN: 1992 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...