3-Brom-1,1-dimetoksypropan ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Bromo-1,1-dimethoxypropane

B1901-25G B1901-5G
TCIAB1901-25GEA 7200 NOK
TCIAB1901-25G TCIAB1901-5G
3-Brom-1,1-dimetoksypropan ≥98.0% (by GC)
3-Brom-1,1-dimetoksypropan
Formel: BrH₂CCH₂CH(OCH₃)₂
MW: 183,05 g/mol
Kokepunkt: 60°C
Flash punkt: 46°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00013245
CAS-nummer: 36255-44-4
EINECS: 252-936-6
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0334f153c8486e0fca339b4a3fc627cc6c53647b&vid=15b06cd2767691441190c767b5991e3105b81751&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR