3,6-Dibromocarbazole ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3,6-Dibromo-9H-carbazole
D2983-1G
D2983-25G
D2983-5G
TCIAD2983-1GEA
220
NOK
TCIAD2983-1G
TCIAD2983-25G
TCIAD2983-5G
3,6-Dibromocarbazole ≥98.0% (by GC)
3,6-Dibromocarbazole
Formel:
C₁₂H₇Br₂N MW: 325 g/mol |
MDL nummer:
MFCD00004961 CAS-nummer: 6825-20-3 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...