2,4,5-Tribromoimidazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2,4,5-Tribromo-1H-imidazole

T2266-25G T2266-5G
TCIAT2266-25GEA 1350 NOK
TCIAT2266-25G TCIAT2266-5G
2,4,5-Tribromoimidazole ≥98.0% (by HPLC, titration analysis)
2,4,5-Tribromoimidazole
Formel: C₃HBr₃N₂
MW: 304,77 g/mol
Kokepunkt: 359 °C (1013 hPa)
Smeltepunkt: 212°C
MDL nummer: MFCD00005184
CAS-nummer: 2034-22-2
EINECS: 217-997-5
FN: 2811
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR