3-Bromo-4-hydroxybenzonitrile ≥95.0% (by GC)
Leverandør: TCI
Warning
B2037-25G B2037-5G
TCIAB2037-25GEA 2350 NOK
TCIAB2037-25G TCIAB2037-5G
3-Bromo-4-hydroxybenzonitrile ≥95.0% (by GC)
3-Bromo-4-hydroxybenzonitrile
Formel: C₇H₄BrNO
MW: 198,02 g/mol
Kokepunkt: 406 °C (1013 hPa)
Smeltepunkt: 155 °C
MDL nummer: MFCD00143096
CAS-nummer: 2315-86-8
EINECS: 219-022-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=273f40387887294f1c84de16645e10690b211820&vid=273f40387887294f1c84de16645e10690b211820&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 154.0 to 161.0 deg-C
Solubility in Methanol almost transparency