2-Amino-4-bromopyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Bromo-2-pyridylamine, 4-Bromopyridin-2-amine
A2339-1G
A2339-5G
TCIAA2339-1GEA
310
NOK
TCIAA2339-1G
TCIAA2339-5G
2-Amino-4-bromopyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-bromopyridine
Formel:
C₅H₅BrN₂ MW: 173,01 g/mol |
MDL nummer:
MFCD01646115 CAS-nummer: 84249-14-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...