4'-Chloro-3'-nitroacetophenone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Chloro-3-nitroacetophenone

C2321-25G C2321-5G
TCIAC2321-25GEA 3275 NOK
TCIAC2321-25G TCIAC2321-5G
4'-Chloro-3'-nitroacetophenone ≥98.0% (by GC)
4'-Chloro-3'-nitroacetophenone
Formel: C₈H₆ClNO₃
MW: 199,59 g/mol
MDL nummer: MFCD00007083
CAS-nummer: 5465-65-6
EINECS: 226-769-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c19102fe19dc954de4dec0741486a50a4bb1049&vid=3c19102fe19dc954de4dec0741486a50a4bb1049&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR