2-Chloro-4-methyl-5-nitropyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Chloro-5-nitro-4-picoline
C2021-1G
C2021-5G
TCIAC2021-1GEA
435
NOK
TCIAC2021-1G
TCIAC2021-5G
2-Chloro-4-methyl-5-nitropyridine ≥98.0% (by GC)
2-Chloro-4-methyl-5-nitropyridine
Formel:
C₆H₅ClN₂O₂ MW: 172,57 g/mol Kokepunkt: 91°C |
MDL nummer:
MFCD00010688 CAS-nummer: 23056-33-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...