N,N-Dimethylbenzamide ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
D0256-25G
D0256-500G
D0256-100G
TCIAD0256-25GEA
405
NOK
TCIAD0256-25G
TCIAD0256-500G
TCIAD0256-100G
N,N-Dimethylbenzamide ≥99.0% (by GC)
N,N-Dimethylbenzamide
Formel:
C₉H₁₁NO MW: 149,19 g/mol Kokepunkt: 256 °C (1013 hPa) Smeltepunkt: 40…42 °C Tetthet: 1,11 g/cm³ (23 °C) Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00008320 CAS-nummer: 611-74-5 EINECS: 210-279-2 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...