(3,3,3-Trifluoropropyl)trimethoxysilane ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Trimethoxy(3,3,3-trifluoropropyl)silane

T2720-25G T2720-5G
TCIAT2720-25GEA 905 NOK
TCIAT2720-25G TCIAT2720-5G
(3,3,3-Trifluoropropyl)trimethoxysilane ≥98.0% (by GC)
(3,3,3-Trifluoropropyl)trimethoxysilane
Formel: F₃CCH₂CH₂Si(OCH₃)₃
MW: 218,25 g/mol
Kokepunkt: 144°C
Tetthet: 1,137 g/cm³ (20 °C)
Flash punkt: 38°C
MDL nummer: MFCD00039266
CAS-nummer: 429-60-7
EINECS: 207-059-3
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f90d64e44fb1ea51624a5097b3715b39566c447&vid=9f90d64e44fb1ea51624a5097b3715b39566c447&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR