2-Aminobenzimidazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole
A0850-25G A0850-5G
TCIAA0850-25GEA 865 NOK
TCIAA0850-25G TCIAA0850-5G
2-Aminobenzimidazole ≥98.0% (by HPLC, titration analysis)
2-Aminobenzimidazole
Formel: C₇H₇N₃
MW: 133,15 g/mol
Kokepunkt: 381 °C (1013 hPa)
Smeltepunkt: 226…231 °C
MDL nummer: MFCD00005596
CAS-nummer: 934-32-7
EINECS: 213-280-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f32f55e12c418aa5b7b4b046b14bed0b600f087&vid=7f32f55e12c418aa5b7b4b046b14bed0b600f087&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 229.0 to 234.0 deg-C
Solubility in Methanol almost transparency