6-Undecanone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Dipentyl ketone, Diamyl ketone, Undecan-6-one
U0043-25ML
U0043-100ML
TCIAU0043-25MLEA
660
NOK
TCIAU0043-25ML
TCIAU0043-100ML
6-Undecanone ≥98.0% (by GC)
6-Undecanone
Formel:
C₁₁H₂₂O MW: 170,3 g/mol Kokepunkt: 226°C Smeltepunkt: 15,5 °C Tetthet: 0,831 g/cm³ (20 °C) Flash punkt: 88°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00009516 CAS-nummer: 927-49-1 EINECS: 213-150-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...