8-Pentadecanone ≥99.0% (by GC)

Leverandør: TCI

Synonymer: Diheptyl ketone, Caprylone

D1545-25G D1545-5G
TCIAD1545-25GEA 3575 NOK
TCIAD1545-25G TCIAD1545-5G
8-Pentadecanone ≥99.0% (by GC)
8-Pentadecanone
Formel: C₁₅H₃₀O
MW: 226,4 g/mol
Kokepunkt: 140°C
Smeltepunkt: 43 °C
Tetthet: 0,787 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009555
CAS-nummer: 818-23-5
EINECS: 212-450-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea2ba9e2520d049e869a80999c107f368c7fcfc4&vid=699075ec0924be436a2476b40c3ad6d2a8bdc8d7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 42.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR