Acetonylaceton ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Hexane-2,5-dione, 2,5-Hexanedione
H0101-25ML
H0101-500ML
H0101-100ML
TCIAH0101-25MLEA
230
NOK
TCIAH0101-25ML
TCIAH0101-500ML
TCIAH0101-100ML
Acetonylaceton ≥97.0% (by GC)
Acetonylaceton
Formel:
C₆H₁₀O₂ MW: 114,14 g/mol Kokepunkt: 188°C Smeltepunkt: -9°C Tetthet: 0,971 g/cm³ (20 °C) Flash punkt: 79°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00008792 CAS-nummer: 110-13-4 EINECS: 203-738-3 Merck Indeks: 14,00071 |
Spesifikasjon Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...