sec-Butyl acetat ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 1-Methylpropyl acetate

A0025-25ML A0025-500ML
TCIAA0025-25MLEA 220 NOK
TCIAA0025-25ML TCIAA0025-500ML
sec-Butyl acetat ≥98.0% (by GC)
sec-Butyl acetat
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokepunkt: 112°C
Smeltepunkt: –98,9 °C
Tetthet: 0,87 g/cm³ (20 °C)
Flash punkt: 18°C
MDL nummer: MFCD00009328
CAS-nummer: 105-46-4
EINECS: 203-300-1
FN: 1123
ADR: 3,II
Merck Indeks: 14,01536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2fa2b8a5b7d2fd4e664a33e140741ac94bfd7db&vid=398d183458e25ab2f77ea771eadbc4848a5df897&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR